Multiwell cell culture plates, Falcon®
Supplier: Corning
736-2026EA 189.78 EUR
736-2026 736-2025 391-0006 734-0949 734-0948 734-0947 734-0028 734-0058 734-0956 734-0019 734-0020 734-0056 734-0055 734-0054
Multiwell cell culture plates, Falcon®
Microplates Cell Culture Plates
Standard tissue culture (TC) and Corning® Primaria™ surface treatments is available. These plates are provided with various colours to suit various detection methods.
  • Crystal-grade virgin polystyrene
  • Nonpyrogenic
  • Sterilised by gamma irradiation
  • Growth area and volume are nominal
Individual and ready-stack (RS) trays are PET (Code 1) and recyclable.
Packaging: Convenient, peel-open medical-style packaging for individual product presentation.