4-well in vitro fertilisation plates, Falcon®
Supplier: Corning
Falcon®
734-0069EA 561.72 EUR
734-0069
4-well in vitro fertilisation plates, Falcon®
Culture Dishes Bioassay Dishes
Falcon® 4-well IVF plates, made from crystal grade PS, sterile, with sliding lid, are pre-tested and certified for in vitro fertilisation (IVF) and designed for consistency and ease of use.
  • Non-embryotoxic, non-pyrogenic and non-cytotoxic
  • Tissue culture treated for a consistent hydrophilic surface
  • Sterilised by gamma irradiation
  • Packaged in peel-open, medical-style packaging
  • Multi-unit bags have reseal tabs
The plates have perfectly flat, optically clear surfaces for optimum manipulation and observation of ova and embryos. Lids are designed for aseptic manipulation and consistent venting to maintain humidification.