Culture plates, Corning® Primaria™
Supplier: Corning
Corning® Primaria™
734-0079EA 302.02 EUR
734-0079 734-0078 734-0077
Culture plates, Corning® Primaria™
Microplates Cell Culture Plates
Unique nitrogen-containing tissue culture surface chemistry. Improves attachment, spreading, and growth for many primary cells or cell lines. Crystal-grade polystyrene modified by proprietary vacuum-gas plasma treatment process.
  • Stable, permanent surface modification
  • Optically clear
  • No special storage required
  • Samples from each lot of Corning Primaria products are analyzed by Electron Scanning for Chemical Analysis (ESCA)
  • Packaged in red color-coded, peel-open, medical-style packages
  • Sterilized by gamma irradiation
  • Nonpyrogenic tested to less than 0,1 EU/ml