Human collagen III (from placenta)
Supplier: Corning Life Sciences
Corning®
734-0104EA 386.33 EUR
734-0104
Human collagen III (from placenta)
Proteins and Peptides
Collagen III is found in several stromal connective tissues including the dermis of young organisms, human skin, and cornea. It can be used as a thin coating on tissue culture surfaces to promote cell attachment and to modulate cell behaviour.
Quality controlled by SDS-PAGE and tested and found negative for bacteria, fungi, and mycoplasma, Source material tested for hepatitis B antigen and HIV-1 antibody