PS, coated with mouse laminin. Laminin, a major structural component of basement membranes, has many varied functions that are mediated by binding to various components of the basement membrane (for example, collagen IV) and to cell-surface receptors. Laminin-coated cell culture plates have a number of applications, including the promotion of cell adhesion, proliferation and differentiation of a variety of cell types, particularly neurons, epithelial cells, myocytes and myoblasts.
Laminin source: Engelbreth-Holm-Swarm (EHS) mouse tumor. Tested for ability to initiate differentiation (neurite outgrowth) of NG-108 rat glioma/mouse neuroblastoma cells. Tested and found negative for bacteria and fungi.
Cellware is stable for at least three months from date of shipment when stored at 2 to 8 °C (36 to 46 °F). Inserts stable for at least three months at 2 to 8 °C (36 to 46 °F). Do not freeze.
Ordering information: Inserts are packaged six per 6-well plate and 12 per 24-well plate.