Fibronectin culture dishes, Corning® BioCoat™
Supplier: Corning
734-0137EA 249.74 EUR
734-0137 734-0111
Fibronectin culture dishes, Corning® BioCoat™
Culture Dishes Cell Culture Dishes
PS coated with a uniform application of human fibronectin. Promotes cell attachment, spreading, proliferation and diffentiation of many cell types, particularly fibroblasts and other mesenchymally-derived cells.
Manufactured in a highly controlled environment and rigorously tested to assure product consistency and performance.