Control cell culture inserts, PET membrane, Corning® BioCoat™
Supplier: Corning
Corning® BioCoat®
734-0215EA 311.38 EUR
734-0215 734-0221
Control cell culture inserts, PET membrane, Corning® BioCoat™
Microplates Insert Plates and Inserts
Corning BioCoat cell culture inserts are pre-coated with extracellular matrix proteins for applications requiring a protein-coated cell surface, such as cell adhesion, growth, invasion, migration and/or differentiation.
Coatings include Corning Matrigel® matrix, fibronectin, collagen and laminin.