Poly-L-lysine and poly-D-lysine cell culture plates, Corning® BioCoat™
Supplier: Corning Life Sciences
Corning® BioCoat™
734-1128EA 2501.1 EUR
734-1128 734-0176 734-0282 734-0316 734-0326 734-0174 734-1121 734-0260 734-0257 734-0317 734-0237 734-0328 734-0300 734-1124 734-0321 734-0151 734-0325 734-0299 734-0258 734-1122 392-0318 734-0278 734-1123 734-0259 734-0250 734-0279 734-0245 734-0286 734-0175 734-0298 734-0121 734-0120 392-0317 734-0327 734-0146
Poly-L-lysine and poly-D-lysine cell culture plates, Corning® BioCoat™
Microplates Cell Culture Plates Coated Plates
PS, coated with poly-L-Lysine or poly-D-Lysine. Poly-D-lysine (PDL) and poly-L-lysine (PLL) are synthetic compounds that enhance cell adhesion and protein absorption by altering surface charges on the culture substrate. PDL and PLL surface treatments support applications including attachment and spreading of a variety of cell lines; cell differentiation and neurite outgrowth; attachment of transfected cell lines; and survival of primary neurons in culture. As PDL and PLL are synthetic molecules, they do not stimulate biological activity in the cells cultured on them, and they do not introduce impurities carried by natural polymers.