Corning® Cell-Tak™ Cell and tissue adhesive
Supplier: Corning
734-1083EA 1226.94 EUR
734-1083 734-1081
Corning® Cell-Tak™ Cell and tissue adhesive
Cell Culture Supplements and Additives
Corning® Cell-Tak™ Cell and Tissue Adhesive is a formulation of polyphenolic proteins extracted from Mytilus edulis (marine mussel). These proteins are the key components of the glue secreted by the mussel to anchor itself to solid substrates in the marine environment. Corning® Cell-Tak™ is used to attach cells or tissue sections to many types of surfaces, including plastic, glass, metal, PTFE, and biological materials. It can simplify the manipulation of biological samples for a number of in vitro techniques, including in situ hybridisation, immunoassays, microinjection, immunohistochemistry, and establishing primary cells in culture. Corning® Cell-Tak™ is biocompatible and demonstrates no species specificity.