Amine and carboxyl microplates, Corning® PureCoat™
Supplier: Corning
Corning® PureCoat™
734-1478EA 1288.12 EUR
734-1478 734-1477 734-1476 734-1475
Amine and carboxyl microplates, Corning® PureCoat™
Microplates Cell Culture Plates
Corning PureCoat Amine (positively charged) and Carboxyl (negatively charged) surfaces provide improved cell attachment, faster cell proliferation, and enhanced recovery post-thaw over standard TC surfaces. These surfaces function with a broad range of primary, transfected, transformed, and fastidious cell types, and have demonstrated utility in serum-reduced or serum-free conditions.