Standard polystyrene microplates 96-well, Corning® Stripwell™
Supplier: Corning
Corning® Stripwell®
734-1515EA 604.38 EUR
734-1515 734-1514 732-5504 732-5548 732-5547 732-5569 CORN9102BC 732-5546 732-5545
Standard polystyrene microplates 96-well, Corning® Stripwell™
Microplates Immunology Microplates and Strips
Corning® Stripwell™ microplates may be used in a variety of laboratory assays. The flat bottom strips are designed to easily break apart and are pre-assembled in an 'egg-crate' style strip holder that allows each individual well to be positioned back into the microplate once broken.
  • Stripwell microplates have 96-well flat bottom polystyrene format
  • Low volume and standard Stripwell microplates have well volumes of 190 µl and 360 µl, respectively
  • 1×8 strips are designed to fit only one way into the strip holder, eliminating the chance of misorientation