Cell culture plates, 96-well
Supplier: Corning
Corning® CellBIND®
734-1798EA 220.08 EUR
734-1798 734-1797 734-1799 734-1546 734-1794 734-1789 734-1796 734-1585 734-1793 392-0291 734-4058 444-1020
Cell culture plates, 96-well
Microplates Cell Culture Plates Standard Plates
PS, clear, with lid.
  • Corning® CellBIND® surface increases wetability for more even and consistent cell attachment
  • Ultra-Low Attachment plates feature a covalently bound hydrogel layer that minimises cell attachment, protein absorption and cellular activation
  • Sterilised by gamma irradiation
  • Certified non-pyrogenic
Recommended working volume: 75 to 200 µl