Multiple well cell culture plates, Costar®
Supplier: Corning
Costar®
392-0214EA 324.45 EUR
392-0214 392-0215 392-0213 734-1607 734-1606 734-1599 734-1598 734-1605 734-1604 734-1582 734-1597 734-1212 734-1211 734-1596 734-1584 734-4067 734-1210
Multiple well cell culture plates, Costar®
Microplates Cell Culture Plates
Clear PS, flat bottom, with lid. Available with a choice of surface treatments.
  • Lids with condensation rings to reduce contamination
  • Uniform footprint for ease of stacking
  • Individual alphanumeric codes for well identification
  • Sterilised by gamma radiation
  • Certified non-pyrogenic