Cell culture roller bottles, Corning®
Supplier: Corning
Corning®
392-0286EA 696.19 EUR
CORN430849_COS 392-0286 734-4190 734-4191 734-1729 392-0307 CORN430195_COS 734-1898 734-1720 734-1884 734-1730 734-1714 734-1702 734-0096 734-0095 734-1870 734-4189
Cell culture roller bottles, Corning®
Bottles Roller Bottles
Virgin PS, sterile.
  • One piece seamless construction
  • All bottles have printed lot numbers to aid product traceability
  • Sterilised by gamma radiation
  • Certified non-pyrogenic
  • Ribbed design provides twice the surface area with the same exterior dimensions
Roller bottles available with smooth or ribbed surface design for expanded culture area. All bottles are treated for optimal cell attachment. The Corning® CellBIND® surface enhances cell attachment under difficult conditions, such as reduced-serum or serum-free medium, resulting in higher cell yields. All bottles are graduated, unless otherwise stated.
Note: Corning recommends 0,2 to 0,3 ml of medium per cm² of growth area, and setting roller rack speeds to provide 0,5 to 1,0 min⁻¹.
Caution: The product(s) on this page are outside the scope of the EU Directive 2019/904