Bottle top vacuum filtration systems, Corning®
Supplier: Corning
Corning®
734-1831EA 108.98 EUR
734-1831 734-1830
Bottle top vacuum filtration systems, Corning®
Filters Bottle Top Filters
Cellulose acetate membrane, PS funnel and PP collection tube.
  • 50 mm diameter membrane
  • Minimises unnecessary transfers by filtering directly into 50 ml centrifuge tube
  • Includes two centrifuge tube stands with each case
  • Each PP centrifuge tube is supplied with an individually wrapped cap for storage
  • Individually packaged, sterile, certified non pyrogenic