Bottle top vacuum filtration systems, Corning®
Supplier: Corning
Corning®
525-3409EA 138.31 EUR
525-3409 734-1848 734-1843 734-1810 734-1842 734-1841 734-1896 734-1811 513-3360 734-1890 513-3361 513-3362 513-3344 513-3345 734-1891
Bottle top vacuum filtration systems, Corning®
Filters Bottle Top Filters
PS bottle top vacuum filters with adapters colour coded by membrane type: Polyethersulfone (PES) yellow; cellulose acetate (CA) orange; nylon (NY) red.
  • Available in 33 mm and 45 mm neck sizes to fit most glass and plastic media storage bottles
  • Individually packaged, sterile and certified non pyrogenic
  • 45 mm neck sizes fit on Corning® plastic storage bottles
Square membrane funnel design provides improved flow rates. Availablefor 150 and 500 ml bottles.
Sterile tissue culture filters in polystyrene funnels attach securely by threaded polyethylene ring to standard media bottles.
Angled hose adapter rotates for easy vacuum line attachment
PES filters give lowest protein binding and fastest flow rates
Ordering information: Complete with filter, dust cover, and polypropylene tubing connector. Media bottle not included.