Chamber slides, Nunc™ Lab-Tek™
Supplier: Thermo Fisher Scientific
Nunc™ Lab-Tek™
734-2060EA 279 EUR
734-2060 734-2062 734-2119 734-2126 734-2127 734-2766 734-2122 734-2056 734-2124 734-2058 734-2125 734-2120 734-2121
Chamber slides, Nunc™ Lab-Tek™
Slides Chamber Slides
PS medium chamber on glass or Permanox™ plastic slide.
  • Cells grow on a standard microscope slide
  • No cell transfer needed prior to visualisation/staining
  • Upper chamber can be removed when culturing is complete
Suitable for use with fluorescent labels.