Glass microfibre prefilters
Supplier: Corning
Corning®
734-2298EA 81.17 EUR
734-2298 734-2299 734-2300 734-2297
Glass microfibre prefilters
Filters Glass Fibre Filters
Glass fiber filters are used a depth filter for prefiltration of solutions.
They have very high particle loading capacity and are ideal for prefiltering dirty solutions and difficult to filter biological fluids such as sera.