VWR®, Cell Culture Flasks, Non Treated or Treated
Supplier: VWR
VWR®
734-3419EA 219 EUR
734-3419 734-2309 734-2804 734-2786 734-2787 734-2600 734-2601 734-2788 734-2789 734-2783 734-2784 734-2785 734-3252 734-3253 734-2809 734-2805 734-2806 734-2807 734-2808 734-2812 734-2813 734-2313 734-2314 734-2810 734-2315 734-2316 734-2811 734-3266 734-3420 734-2310 734-2311 734-2312 734-2790 734-2791 734-2792 734-3265
VWR®, Cell Culture Flasks, Non Treated or Treated
Flasks Cell Culture Flasks
These cell and tissue culture flasks made of PS are perfect for cell growth.
  • Available with five different growth areas
  • Flask surface is flat and free from striations to maximise usable growth area
  • Two different HDPE cap styles can be used in both open and closed systems
  • Innovative angled neck design offers good pipette and cell scraper access
  • Triangular top with wide base provides stability
  • Easy stacking
  • Frosted area near the neck for easy marking/writing
  • Engraved graduations on multiple sides
  • 100% integrity tested
  • Sterilised by electron beam irradiation
  • Free from DNase and RNase and non pyrogenic
Non treated: Cell and tissue culture flasks are ideal for applications where cell attachment is not desired.
TC treated: For optimal cell attachment and growth. TC treated, extended (*): Deeper flask for increased recommended working volume.
Increased cell attachment surface: This highly hydrophilic surface offers a significant advantage over the traditional cell culture surface. The surface treatment can improve cell spreading and attachment, and is suitable for cells that may adhere poorly due to cell phenotype or stressful culture conditions and for those which normally require additional biological coatings for attachment.
734-3419 and 734-3420 perfectly fits for Thermo Fisher incubators. Three can be placed in parallel on each floor to fully use the space.
Certifications: ISO 13485:2003 and ISO 9001:2008 certified.