VWR® Bottle Assemblies, PETG, Sterile, Single-Use
Supplier: VWR
VWR®
734-2845EA 948 EUR
734-2845 VWRU10830-298 734-2846 VWRU10830-296 VWRU10830-294 734-2844 VWRU75835-988 734-2850 734-2894 734-2851 VWRU10830-300
VWR® Bottle Assemblies, PETG, Sterile, Single-Use
Bottles Cell Culture Bottles
Sterile PETG containers, with weldable tubings, provide the maximum amount of flexibility and reliability. These vessels are assembled in an ISO Class 7 cleanroom and are manufactured from USP Class VI materials. They come in a comprehensive size selection with tubing that is completely weldable/sealable.
  • Sterile bottles come individually bagged and process ready
  • Multiple bottle sizes allow for process-specific flexibility
  • When used with sterile VWR tube sets, custom possibilities are endless
  • Gamma irradiated to Sterility Assurance Level (SAL) 10⁻⁶
Assemblies have been designed with a broad spectrum of applications in mind, including life sciences and general lab use, environmental assays, also production and cleanroom applications. All product contact materials are animal-free or processed in accordance to EMEA/410/01. The assemblies are able to withstand temperatures from –20 to +65 °C without losing integrity. All assemblies are individually double-bagged with product label located on the inner bag. Gamma irradiated to Sterility Assurance Level (SAL) 10-6.
Caution: *The product(s) on this page are outside the scope of the EU Directive 2019/904.