Bottle assemblies, PC, sterile, single-use, VWR®
Supplier: VWR, part of Avantor
VWR®
734-2853EA 835 EUR
734-2853 734-2854 734-2895 734-2855 734-2852 734-2856
Bottle assemblies, PC, sterile, single-use, VWR®
Bottles Cell Culture Bottles
Sterile polycarbonate containers, with weldable tubing, provide the maximum amount of flexibility and reliability. These vessels are assembled in an ISO Class 7 cleanroom and are manufactured from USP Class VI materials. They comes in a comprehensive size selection with tubing that is completely weldable/sealable.
  • Sterile bottles come individually bagged and process ready
  • Multiple bottle sizes allow for process-specific flexibility
  • When used with sterile VWR® tube sets, custom possibilities are endless
  • Gamma irradiated to Sterility Assurance Level (SAL) 10-6
Assemblies have been designed with a broad spectrum of applications in mind, including life sciences and general lab use, environmental assays, also production and cleanroom applications. All product contact materials are animal-free or processed in accordance to EMEA/410/01. The assemblies are able to withstand temperatures from –20 to +65 °C without losing integrity. All assemblies are individually double-bagged with product label located on the inner bag. Gamma irradiated to Sterility Assurance Level (SAL) 10-6.
Caution: *The product(s) on this page are outside the scope of the EU Directive 2019/904