96-Well clear polystyrene microplates, Corning®
Supplier: Corning
Corning®
734-1629EA 604.94 EUR
734-1629 734-1628 734-1799 734-1798 734-1611 734-1797 734-1796 734-1554 734-1659 732-5521 732-5543 734-1658 734-1657 734-1791 444-1020 734-1790 732-5568 732-5567 734-1553 734-1795 734-1794 734-1793 734-2291 CORN3628BC 734-1546 734-1645 734-1644 734-1786 734-1785 734-1627 734-1548 734-1789 734-1624 734-4058 734-1784 734-1641 734-1585 734-1783
96-Well clear polystyrene microplates, Corning®
Microplates Cell Culture Plates
Cell culture microplates are sterile and nonpyrogenic. Lids available where indicated.
Cat. nos. 3798 and 3898 processed to improve hydrophilicity for hemagglutination and similar assays. Cat. Nos. 3585 and 3595 have special low evaporation lids. Cat. No. 3841 is aseptically manufactured.