384-well clear polystyrene microplates, Corning®
Supplier: Corning
Corning®
732-5520EA 1144.93 EUR
732-5520 732-5537 732-5536 734-2294 734-4121 732-5539 732-5538
384-well clear polystyrene microplates, Corning®
Microplates Cell Culture Plates
PS, flat bottom.
  • Total well volume of 112 μl; working well volume of 20 to 80 μl
  • Cell culture plates sterilised by gamma radiation and certified non pyrogenic
  • Universal optics NBS™ plate manufactured using an advanced polymer with high clarity and improved chemical resistance
  • Lids supplied with sterile plates, other lids can be ordered separately
White walled plates enhance luminescent signals and have low background luminescence and fluorescence. Black walled plates have low background fluorescence, minimal light scatter.