96-well solid black and white polystyrene microplates, Corning®
Supplier: Corning
Corning®
734-4171EA 324.15 EUR
734-4171 734-4170 CORN4591 734-1623 734-1667 734-1622 734-1666 734-1665 734-1643 CORN3789A 734-1664 732-5522 734-4117 734-1626 734-1549 734-1625 732-5596 732-5515 734-4169 734-1663 734-4153 734-1662 732-5517
96-well solid black and white polystyrene microplates, Corning®
Microplates Cell Culture Plates
These 96-well solid black and white polystyrene microplates are designed to reduce well-to-well cross-talk.
White microplates enhance luminescent signals and have low background luminescence.
Black microplates have low background fluorescence and minimise light scattering.