Cell culture storage bottles, square, PC
Supplier: Corning
734-4208EA 186.51 EUR
734-4208 734-4207 734-4205 734-4206
Cell culture storage bottles, square, PC
Bottles Cell Culture Bottles
PC, square.
  • More break-resistant than other glass or plastic bottles
  • Screened white enamel graduations are easier to see than moulded graduations
  • Large white marking spot for easier identification
  • Bottles can be autoclaved once at 121 °C and 15 psi (repeated autoclaving weakens PC and is not recommended)
  • Can be stored at –80 °C
These storage bottles are easier to handle, require less space on the shelf or in the autoclave, and are ideal for mixing, sampling and storage.