Adhesive seals
Supplier: Greiner bio-one
BREATHseal™ EASYseal™
GREI676001BEEA 0 EUR
GREI676001BE GREI676050_100 GREI676002_200 736-0127 736-0129
Adhesive seals
Seals Microplate Seals
These adhesive seals offer an alternative to heat seals or sealing mats. EASYseal™ is a transparent adhesive film designed to cover microplates to protect samples from evaporation and contamination. It is suitable for routine laboratory use, including ELISA applications. BREATHseal™ is a gas permeable, pierceable membrane coated with an acrylic adhesive. It is ideal for bacterial, yeast and cell culture applications, assuring the protection of cell contents from contamination whilst maintaining oxygenation.
  • Free from RNase, DNase and human DNA
  • Non pyrogenic