Deep well plates, 96- and 48- well, Axygen®
Supplier: Corning Life Sciences
Axygen®
736-0345EA 186 EUR
736-0345 738-0187 738-0185 737-0159 736-0336 736-0358 736-0349 738-0186 736-0339 736-0335 736-0344 736-0359 736-0348 738-0188
Deep well plates, 96- and 48- well, Axygen®
Microplates Deep Well Microplates
PP. Used for sample collection, cell culture and long term storage.
  • Excellent chemical resistance and temperature resistance down to −80 °C
  • Can withstand centrifugation up to 4000×g
Two types of sealing mats are suitable for use with these plates:
AxyMat™ sealing mats are designed for high throughput screening and storage applications. Made of silicone, they can be autoclaved, and following a bleach wash and ethanol rinse protocol, they can also be re-used. AxyMat™ sealing mats are pierceable and self-sealing, ideal for automated processes, and can be used in sub-zero environments.
ImpermaMat™ sealing mats are chemically resistant. They are made for use in assays involving strong solvents such as DMSO and similarly harsh solvents. Re-usable. They feature a tight seal to the plate so there is no sample evaporation or well-to-well evaporation. Usable within a wide temperature range: from −80 up to +121 °C.