Deep well and assay plates, Axygen®
Supplier: Corning
Axygen®
736-0332EA 25.8 EUR
736-0332 736-0344 736-0345 736-0339 736-0357 736-0335 736-0336 736-0358 738-0186 736-0359 738-1000 736-0348 738-0187 736-0349 738-0185 732-1371 732-1370 732-1373 732-1372 737-0159
Deep well and assay plates, Axygen®
Microplates Deep Well Microplates
Axygen® deep well and assay plates are used for sample collection, in vitro growth chambers and long-term storage.
  • 0,6 ml deep well plate
  • ANSI/SLAS 1-2004: Microplates - footprint dimensions
  • Ultra-flat surface ensures proper sealing with heat sealing film
Made of polypropylene, they offer excellent chemical resistance and will withstand temperatures down to −80 °C.