Microplate washer, Wellwash™
Supplier: Thermo Fisher Scientific
Wellwash™
736-0422EA 5890 EUR
736-0422 736-3008
Microplate washer, Wellwash™
Microplate Washers
Wash 96-well plates securely with the Thermo Scientific™ Wellwash™ Microplate Washer, an easy-to-use, reliable microplate-strip washer for routine ELISA applications. The Wellwash features a graphical user interface, local language versions and USB port for optimal ease-of-use and convenience.
  • Large color clear display and keypad for easy, convenient use
  • User interface is logic and intuitive for quick protocols development
  • Easy-to-use software requires minimal training for fast startup
  • Unpressurized bottles are safe and secure to use
  • Liquid level sensors are present in both wash and waste bottle for security
  • Automatic prime feature assure safe performance
  • Aerosol cover prevents aerosolization of infectous diseases
  • Plate sensor recognizes if plate is present
  • Wellwash is designed to efficiently remove the excess liquid from the wells
  • Low residual volumes provide optimal washing performance and reliable assay results
Recommended for ELISA-based assays.
Ordering information: One-year warranty.
Delivery information: Includes 1×2 L wash bottle, 1×2 L waste bottle, 1×8 or 1×12 wash head, aerosol cover.