SpectraMax ABS Plus UV/Vis Single-Mode Microplate Reader with built-in Cuvette Port. Including 12 months parts and labor warranty and 1 additional year of MD Service Plan covering parts and labor. System Includes: Tunable Monochromator-based Spectral 1 * 1 items
Supplier: Molecular Devices
Microplate Reader, Mode of operation: Absorbance, SpectraMax® ABS PLUS UV/Vis absorbance microplate reader, single-mode, with current version of SoftMax® Pro and 1 additional year MD service plan covering parts and labour, Plate types: 96- to 384-well plates and cuvettes, Wavelength range: 190 - 1000 nm, : N/A, Measurement time: Normal mode: 96-well plate in 9 s and 384-well plate in 29 s;: Speed read mode: 96-well plate in 5 s and 384-well plate in 16 s (minimum times), : N/A, Incubation range: 5 °C above ambient to 45 °C
Learn more
About VWR
Inquire for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Dual use goods can only be delivered within the European Union.
Dual use goods can only be delivered within the European Union.
This product has been blocked by your organization. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
This product is no longer available. Alternatives may be available by searching with the VWR Catalog Number listed above. If you need further assistance, please call VWR Customer Service at +43 1 97002 - 0.