96-Well cell culture plates, Falcon®
Supplier: Corning
Falcon®
734-2480EA 332.76 EUR
734-2480 736-2027 734-0080 732-2195 734-0027 734-0025 732-2194 734-0955 734-0954 734-0057 734-0023 734-1376 734-0396
96-Well cell culture plates, Falcon®
Microplates Cell Culture Plates
Falcon® 96-well microplates are available in a variety of configurations and surface chemistries to meet a wide range of applications with fluorescence, luminescence, radiometric or colorimetric endpoints.
  • Flat bottom, with lid
  • Black wells with clear bottom
  • Working volume: 25 to 340 µl
  • Convenient, peel-open packaging
  • Available with untreated or tissue culture-treated surface chemistries
  • Individual and Ready-Stack (RS) trays are PET (Code 1) and recyclable
Corning® Primaria™ tissue culture treatment additionally incorporates nitrogen-containing functional groups that have been shown to improve attachment and spreading of some cell types.
Falcon 96, 384, and 1536 well microplates are compatible with automation systems and meet standard ANSI/SBS footprint dimensions for microplates.