Immunology tubes
Supplier: Greiner bio-one
MICROLON®
738-0003EA 184 EUR
738-0003 738-0001 738-0002 738-0000
Immunology tubes
Tubes Immunology Tubes
PS, clear. Star bottomed tubes have an increased surface area and thus make it possible to bind larger amounts of antigens or antibodies.
Immuno tubes are often used for determining hormone levels, for example TSH (thyroid stimulating hormone).
Available with either MICROLON® 200 (medium binding) and MICROLON® 600 (high binding) surfaces.
Ø×L: 12×75 mm