Clear bottom, black and white, polystyrene microplates, 384-well, Corning®
Supplier: Corning
Corning®
CORN3765EA 1796.32 EUR
CORN3765 CORN3764 CORN3763 734-4086 CORN3762 CORN3769 CORN4588 CORN3768 CORN3767 CORN3766 CORN4596 738-0210 738-0211 CORN4690 CORN3764BC 734-4168 734-4087 734-1200 734-4088
Clear bottom, black and white, polystyrene microplates, 384-well, Corning®
Microplates HTS Microplates
PS, flat bottom. Suitable for fluorescent and luminescent assays using either top or bottom detection plate readers.