Micro particles based on polystyrene, analytical standard, Supelco®
Supplier: Merck
84135-10ML-FEA 520 EUR
84135-10ML-F 79807-5ML-F 74964-10ML-F 74491-10ML-F 74964-5ML-F 84135-5ML-F 74491-5ML-F
Micro particles based on polystyrene, analytical standard, Supelco®
Standards Particle Size Standards
For every lot exact values of particle size and standard deviation are determined with an accuracy of 0,01 μm using a Coulter Multisizer.