Gel drying solution for protein gels
Supplier: G-Biosciences
786-686EA 569 EUR
786-686 786-684 786-685
Gel drying solution for protein gels
Electrophoresis Gels Precast Gels
This gel drying solution effectively and evenly dries protein electrophoresis gels by the mechanism of passive evaporation and is suitable for permanent gel storage and densitometry.
  • Ready to use
  • Regulates rate of drying
  • Prevents cracking of the gel
  • Compatible with all drying membranes