Disposable spin columns for protein purification
Supplier: G-Biosciences
786-726EA 64.7 EUR
786-726 786-724 786-720
Disposable spin columns for protein purification
Filters Centrifugal Filters
These disposable spin columns are designed for protein purification. These spin columns are uniquely designed, with a snap off end converting to a closure for the column for easy manipulation and use.
These columns can be spun with the stopper attached and then snapped off to elute solutions. For further storage or incubation, simply push the column into the stopper.