Beakers, tall form, DURAN®
Supplier: DWK Life Sciences
801-3141EA 12.7 EUR
801-3141 213-1190 801-3129 801-3136 215-1012 213-0253 801-3154
Beakers, tall form, DURAN®
Beakers
DURAN®, borosilicate glass 3.3, clear, without spout.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion, giving relatively high resistance to temperature changes
Graduated for approximate reading of contents and labelling area.
With printed Retrace Code (batch identification).
Certifications: ISO 3819