VWR® Label Dots on Rolls
Supplier: VWR
VWR®
817-5018EA 223 EUR
817-5018 817-5009 817-5021 817-5022 817-5002 817-5004
VWR® Label Dots on Rolls
Labels
These label dot rolls are made from chemically-inert polyester with a unique adhesive formulated to adhere to laboratory glass and plasticware.
  • Withstand temperatures from –196 to +121 °C
  • No need to laminate
  • Use on tubes, racks and boxes
VWR® labels are ideal for all cryostorage and can also be used in most incubators.