VWR®, Laboratory Notebooks
Supplier: VWR
VWR®
818-0069EA 46.5 EUR
818-0069 818-0068 818-0081 818-0080 818-0283 818-0085 818-0282 818-0084 818-0083 818-0082 818-0089 818-0088 818-0087 818-0086 818-0115 VWRU89005-136 818-0114 818-0113 818-0112 818-0079 VWRU89005-132 VWRU89005-138 818-0070 818-0092 818-0091 818-0090 818-0096 818-0074 818-0095 818-0073 818-0072 818-0094 818-0071 818-0093 818-0078 818-0099 818-0077 818-0076 818-0098 818-0097 818-0075
VWR®, Laboratory Notebooks
Notebooks
Imitation leather cover with coating, for durability in laboratory environments. Acid-free, archive safe paper (90 gsm). Includes laboratory notebook guidelines page, table of contents and user data page. Available in different language formats: English only, English-French, English-German, English-Spanish and English-Italian.
  • Pages are sewn in sections with round binding so that book lays flat
  • Scientific 5 mm grid or ruled format with date and witness signature blocks
  • Waterproof
Format: A4 (21×29,7 cm) or A5 (14,8×21 cm). A4 books available in three sizes: 72, 120 and 216 pages. A5 available with 96 pages.
Colour: blue
All books made in accordance with GLP, GMP and GCP guidelines.