Detergents and cleaning agents, LABWASH® Premium
Supplier: VWR Chemicals
84551.320EA 26.76 EUR
84551.320 84551.365 84556.365 84547.360 84545.410 84556.360 84547.320 84553.410 84555.360 84546.410 84550.410BOM 84555.365 84551.360 84551.440 84546.360 84550.365 84546.320 84549.320 84554.360 84549.365 84547.410 84554.320 84550.360 84557.290 84550.440 84861.410 84550.320 84552.410 84548.290 84556.410 84545.320 84553.360 84551.410 84548.410 84545.360 84549.360 84557.410 84553.320 84553.365 84550.410 84552.360 84549.410 85831.360 84552.365
Detergents and cleaning agents, LABWASH® Premium
Cleaning Agents Detergents and Surface Cleansers
Compact HDPE canisters (Cat. Nos. 845xx.365) are compatible with Miele dosing cabinets.
  • Residue-free laboratory glassware cleaner
  • Excellent cleaning power at low dosage levels
  • NTA-free (no nitrilotriacetic acid)
  • Protects the environment, active ingredients are biodegradable
  • Acid-rinse concentrates available to neutralise alkaline cleaners
pH value at 1% concentration.