VWR®, Water bath stabiliser, AQUA RESIST WB
Supplier: Avantor
VWR®
86174.440EA 152 EUR
86174.440 86174.360
VWR®, Water bath stabiliser, AQUA RESIST WB
Cleaning Agents Water Bath Protection Reagents
A ready to use, liquid product designed to stabilise and protect water baths, cooling circuits, water circuits and incubators.
  • Working temperature up to 70 °C
  • Non corrosive for conventional materials
  • Good wetting characteristics
  • Low foaming
  • Blue dye for visualisation
AQUA RESIST WB is characterised by a specific combination of several ingredients. Surfactants cause stabilisation of the water with simultaneous good wetting. Stabilisers and corrosion inhibitors ensure high material compatibility and thus for a wide range of applications even with longer exposure times. The blue colour makes visual inspection of the existing product possible.