VWR® IPASEPT® 70 Cleaning Solution
Supplier: Avantor
VWR®
93002.9010EA 208 EUR
VWRS93002.0516 93002.9010 VWRS93002.5000 93002.1016
VWR® IPASEPT® 70 Cleaning Solution
Cleaning Agents Detergents and Surface Cleansers
Iso-propyl alcohol Ph. Eur. 70% v/v, with purified water Ph. Eur. 30% v/v, filtered at 0,2 μm.
  • Filled under GMP conditions in cleanroom ISO Class 7
  • Certificate of Analysis available for each batch
For surface spray cleaning, with or without wipes. Also for cleaning gloved hands before entry into cleanrooms. Ideal for wiping down entrances to controlled environments.