Micro particle size standard based on polystyrene monodisperse, analytical standard, Supelco®
Supplier: Merck
87896-5ML-FEA 373 EUR
87896-5ML-F 95531-5ML-F 95585-5ML-F 72938-5ML-F 81494-5ML-F 89756-5ML-F 81494-10ML-F 80177-5ML-F 72938-10ML-F 84192-5ML-F 88511-5ML-F 80304-5ML-F 95585-10ML-F
Micro particle size standard based on polystyrene monodisperse, analytical standard, Supelco®
Standards Particle Size Standards
For every lot exact values of particle size and standard deviation are determined with an accuracy of 0,001 μm.