Glycylglycine (Gly-Gly)

Supplier: Thermo Fisher Scientific
A10523.36EA 512 EUR
A10523.36 A10523.14 A10523.22
Glycylglycine (Gly-Gly)
Proteins and Peptides
Buffer with a useful pH range 7,5 to 8,9

  • EINECS: 209-127-8
  • GHS symbol and signal word: GHS07 and warning
  • Melting point: ca 220 °C (dec.)
  • Merck index: 14,4503
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR