Glycylglycylglycine (Gly-Gly-Gly)

Supplier: Thermo Fisher Scientific
A13778.06EA 76 EUR
A13778.06 A13778.03 A13778.14
Glycylglycylglycine (Gly-Gly-Gly)
Proteins and Peptides
  • EINECS: 209-122-0
  • Melting point: ∼240 °C (dec.)
  • Merck index_V14: 14,6579
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR