DNA ligation kit
Supplier: Agilent Technologies
AGLS203003EA 404 EUR
AGLS203003
DNA ligation kit
Nucleic Acid Reagents Cloning Kits and Ligation
The DNA Ligation Kit includes T4 DNA ligase for highly efficient ligations.
  • For both sticky-end and blunt-end ligations
  • Reagents guaranteed to be DNase free while providing optimal ligation efficiency
  • Includes a plasmid control and a lambda control
The kit is optimised for use with lambda arms and plasmid vectors.
Delivery information: Kit contains T4 DNA ligase (300 U) 10X ligase buffer plasmid control Lambda control 10 mM rATP.
Caution: For research use only. Not for use in diagnostic procedures.