Anti-LAMP2 Rat Monoclonal Antibody

Supplier: Enzo Life Sciences
ALEXADIVAMEN003FEA 742 EUR
ALEXADIVAMEN003F ALEXADIVAMEN003D
Anti-LAMP2 Rat Monoclonal Antibody
Antibodies
Lysosome-associated membrane proteins (LAMP1 and LAMP2) are type I integral membrane protein that are transported from the trans-Golgi network to endosomes and then lysosomes. Following activation of platelets, T-cells, neutrophils, or endothelium, LAMP1 and LAMP2 are expressed on the cell surface. Cell surface LAMP1 and LAMP2 have been shown to promote adhesion of human peripheral blood mononuclear cells (PBMC) to vascular endothelium, and loss of LAMP2 expression is associated with impaired autophagy observed in Danons disease.

  • For Immunocytochemistry, Immunoprecipitation, Western Blot

Recognizes mouse and rabbit LAMP2.

Type: Primary
Antigen: LAMP2
Clonality: Monoclonal
Clone:
Conjugation:
Epitope:
Host: Rat
Isotype: IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR