Chlorpromazine hydrochloride ≥98% (by HPLC)

Supplier: Enzo Life Sciences

ALEXALX270171G005EA 105 EUR
ALEXALX270171G005
Chlorpromazine hydrochloride ≥98% (by HPLC)
Chlorpromazine hydrochloride

Antipsychotic working on a variety of receptors in the CNS


Inhibits calmodulin stimulation of cyclic nucleotide phosphodiesterase. Dopamine antagonist. D2 antagonist. H1 antagonist. Inhibits TNF-alpha production. Potent PLA2 inhibitor. Inhibits nitric oxide synthase (NOS) in mouse brain and prevents lipopolysaccharide induction of NOS in mouse lung. Antipsychotic.

Formula: C₁₇H₁₉ClN₂S
MW: 355,32 g/mol
MDL Number: MFCD00012654
CAS Number: 69-09-0
https://export.vwr.com/stibo/web/std.lang.all/74/83/45347483.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR