Bioactive lipid library plates, SCREEN-WELL®
Supplier: Enzo Life Sciences
ALEXBML-2800-0100EA 7440 EUR
ALEXBML-2800-0100 ALEXBML-2800-0500
Bioactive lipid library plates, SCREEN-WELL®
Microplates HTS Microplates
SCREEN-WELL® bioactive lipid library contains 190 bioactive lipids in a 96 well format. Ideal for screening or identifiying recombinant orphan G protein-coupled receptors, target validation, secondary screening, validating new assays and for routine pharmacological applications.
Lipid library contains 190 compounds including agonists, antagonists, endocannabinoids, farnesyl/geranylgeranyl derivates, HETEs deHETEs, hepoxilins, polyunsaturated fatty acids, leukotrienes, lipoxins, LPA and phosphatidic acids, octadecanoids, PAFs, prostaglandins and thromboxanes, retinoids, vitamin D metabolites, sphingolipids.