Endocannabinoid library plates, SCREEN-WELL®
Supplier: Enzo Life Sciences
ALEXBML-2801-0100EA 3640 EUR
ALEXBML-2801-0100 ALEXBML-2801-0500
Endocannabinoid library plates, SCREEN-WELL®
Microplates HTS Microplates
SCREEN-WELL® Endocannabinoid Library contains 58 compounds with defined, putative, potential or speculative activity at cannabinoid (CB) and vanilloid (VR) receptors.
The library is an array of 10 different fatty acids and 6 different polar head groups. Ligands are supplied dissolved in biocompatible solvents at 1 mM and aliquoted into deep-well plates. The library is a rich source of ligands for receptor de-orphaning and includes the following classes of ligands: Amides, ethanolamides, lipo-amino acids, acyl-GABAs, acyl-dopamines.